Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282062.1 | NP_001268991.1 | 3 | 257 | ||
NM_001282063.1 | NP_001268992.1 | 3 | 219 | |||
NM_001282068.1 | NP_001268997.1 | 2 | 184 | |||
NM_003586.2 | NP_003577.2 | 2 | 218 | |||
XM_011545975.1 | XP_011544277.1 | 4 | 643 | |||
XM_017023776.1 | XP_016879265.1 | 6 | 861 | |||
XM_017023777.1 | XP_016879266.1 | 3 | 798 | |||
GenBank mRNA | BC041769.1 | - | 3 | 220 | 4819032 | |
BC055284.1 | - | 2 | 218 | 5767656 | ||
BC063436.1 | - | 3 | 183 | 5394616 | ||
D31897.1 | - | 2 | 139 |