Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008895.2 | NP_001008895.1 | 19 | 2191 | ||
NM_001278513.1 | NP_001265442.1 | 19 | 2335 | |||
NM_001278514.1 | NP_001265443.1 | 19 | 2003 | |||
NM_003589.2 | NP_003580.1 | 19 | 2017 | |||
XM_011537523.2 | XP_011535825.1 | 18 | 2551 | |||
GenBank mRNA | AB012193.1 | - | 13 | 1365 | ||
AB178950.1 | - | 19 | 2068 | |||
AF077188.1 | - | 19 | 1928 | |||
AK296700.1 | - | 11 | 1312 | |||
AK314835.1 | - | 19 | 2290 | |||
AL833355.1 | - | 18 | 1955 | |||
AY365124.1 | - | 19 | 2068 | |||
BC008308.1 | - | 19 | 2079 | 3537176 | ||
U58090.1 | - | 9 | 1055 |