Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261823.1 | NP_001248752.1 | 7 | 841 | ||
NM_003599.3 | NP_003590.1 | 8 | 932 | |||
NM_181356.2 | NP_852001.1 | 10 | 1117 | |||
XM_011514949.2 | XP_011513251.1 | 10 | 953 | |||
XM_011514950.1 | XP_011513252.1 | 8 | 932 | |||
XM_011514951.2 | XP_011513253.1 | 8 | 4993 | |||
XM_011514952.2 | XP_011513254.1 | 8 | 1979 | |||
XM_011514953.2 | XP_011513255.1 | 8 | 762 | |||
XM_011514954.2 | XP_011513256.1 | 8 | 793 | |||
XM_017011369.1 | XP_016866858.1 | 10 | 5181 | |||
XM_017011370.1 | XP_016866859.1 | 8 | 746 | |||
XM_017011371.1 | XP_016866860.1 | 9 | 4107 | |||
XM_017011372.1 | XP_016866861.1 | 8 | 821 | |||
XM_017011373.1 | XP_016866862.1 | 9 | 874 | |||
XM_017011374.1 | XP_016866863.1 | 7 | 628 | |||
GenBank mRNA | AF069734.1 | - | 7 | 648 | ||
AF073930.1 | - | 8 | 719 | |||
AK303838.1 | - | 7 | 654 | |||
AK313874.1 | - | 8 | 922 | |||
BC035816.1 | - | 8 | 809 | |||
BC050384.1 | - | 10 | 999 | 5758833 |