Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDC42BPA SIRNA Assays ›
Gene Symbol:
CDC42BPA
Gene Name:
CDC42 binding protein kinase alpha
Gene Aliases:
MRCK, MRCKA, PK428
Chromosome Location:
Chr.1: 226989865 - 227318492 on Build GRCh38
UniGene ID:
Hs.35433
Species:
Human
siRNA ID
HSS112353
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDC42BPA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003607.3
NP_003598.2
34
5713
NM_014826.4
NP_055641.3
33
5470
XM_005273317.3
XP_005273374.1
37
6164
XM_005273318.3
XP_005273375.1
37
6155
XM_005273320.3
XP_005273377.1
36
6146
XM_005273321.3
XP_005273378.1
35
6011
XM_005273322.3
XP_005273379.1
35
5930
XM_005273324.3
XP_005273381.1
35
5960
XM_006711834.3
XP_006711897.1
36
6116
XM_006711835.3
XP_006711898.1
36
6059
XM_011544306.2
XP_011542608.1
38
6458
XM_011544307.2
XP_011542609.1
37
6098
XM_011544308.2
XP_011542610.1
37
6017
XM_011544309.2
XP_011542611.1
37
5960
XM_017002578.1
XP_016858067.1
36
5969
XM_017002579.1
XP_016858068.1
39
6616
XM_017002580.1
XP_016858069.1
37
4903
XM_017002581.1
XP_016858070.1
37
6529
GenBank mRNA
AB007920.2
-
19
2563
AB208968.1
-
33
5344
AJ518975.1
-
34
4770
AJ518976.1
-
33
4527
AK027000.1
-
10
1460
AY166586.1
-
33
4527
BC136333.1
-
34
4737
9020320
BX648219.1
-
12
1694
BX648693.1
-
21
2755
CR933723.1
-
21
2755
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
cell migration
actomyosin structure organization
actin cytoskeleton reorganization
intracellular signal transduction
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline