Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIRIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197323.1 | NP_001184252.1 | 5 | 1034 | ||
NM_003609.4 | NP_003600.2 | 6 | 1972 | |||
GenBank mRNA | AJ223349.1 | - | 5 | 528 | ||
AJ223350.1 | - | 5 | 1350 | |||
AJ223351.2 | - | 6 | 1869 | |||
AK075145.1 | - | 5 | 1285 | |||
AK312504.1 | - | 6 | 1488 | |||
BC000588.2 | - | 6 | 1942 | 3345739 |