Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015885.1 | NP_001015885.1 | 8 | 986 | ||
NM_003610.3 | NP_003601.1 | 8 | 962 | |||
XM_005260582.2 | XP_005260639.2 | 8 | 745 | |||
XM_005260583.2 | XP_005260640.1 | 8 | 751 | |||
XM_011529087.2 | XP_011527389.1 | 8 | 745 | |||
XM_011529088.2 | XP_011527390.1 | 8 | 1117 | |||
XM_011529089.1 | XP_011527391.1 | 8 | 878 | |||
XM_017028108.1 | XP_016883597.1 | 8 | 1082 | |||
GenBank mRNA | AK292247.1 | - | 8 | 1018 | ||
AK307966.1 | - | 8 | 825 | |||
AK309069.1 | - | 7 | 771 | |||
AK309743.1 | - | 8 | 798 | |||
AY336091.1 | - | 7 | 639 | |||
AY349350.1 | - | 8 | 986 | |||
BC103754.1 | - | 8 | 803 | 6501538 | ||
BC106923.1 | - | 8 | 760 | 40032941 | ||
BC106924.1 | - | 8 | 760 | 40032942 | ||
U84720.1 | - | 8 | 808 |