Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003611.2 | NP_003602.1 | 17, 18 | 2739 | ||
XM_005274604.2 | XP_005274661.1 | 16, 17 | 2619 | |||
XM_005274606.3 | XP_005274663.1 | 18, 19 | 2881 | |||
XM_011545592.2 | XP_011543894.1 | 18, 19 | 2710 | |||
XM_011545594.2 | XP_011543896.1 | 16, 17 | 2160 | |||
XM_011545595.1 | XP_011543897.1 | 16, 17 | 2170 | |||
XM_011545597.1 | XP_011543899.1 | 18, 19 | 2795 | |||
XM_011545598.1 | XP_011543900.1 | 6, 7 | 1238 | |||
XM_017029909.1 | XP_016885398.1 | 17, 18 | 2315 | |||
XM_017029910.1 | XP_016885399.1 | 18, 19 | 2523 | |||
XM_017029911.1 | XP_016885400.1 | 9, 10 | 1560 | |||
GenBank mRNA | AK225847.1 | - | 16, 17 | 2549 | ||
AK290354.1 | - | 17, 18 | 2724 | |||
AK297104.1 | - | 9, 10 | 1519 | |||
BC096344.3 | - | 16, 17 | 2434 | 40008704 | ||
BC096345.1 | - | 16, 17 | 2448 | |||
GU727634.1 | - | 16, 17 | 2380 | |||
Y15164.1 | - | 17, 18 | 2691 | |||
Y16355.1 | - | 17, 18 | 3355 |