Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195303.2 | NP_001182232.1 | 3 | 569 | ||
NM_001254748.1 | NP_001241677.1 | 6 | 627 | |||
NM_001254749.1 | NP_001241678.1 | 5 | 754 | |||
NM_003617.3 | NP_003608.1 | 5 | 742 | |||
GenBank mRNA | AB008109.1 | - | 5 | 485 | ||
AF030108.1 | - | 5 | 480 | |||
AF159570.1 | - | 5 | 448 | |||
AF493929.1 | - | 5 | 404 | |||
AJ891044.1 | - | 3 | 231 | |||
AK315357.1 | - | 5 | 485 | |||
BC030059.1 | - | 5 | 467 | 4098248 | ||
BU172751.1 | - | 5 | 484 | |||
BX537427.1 | - | 5 | 486 |