Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PKP4 SIRNA Assays ›
Gene Symbol:
PKP4
Gene Name:
plakophilin 4
Gene Aliases:
p0071
Chromosome Location:
Chr.2: 158456880 - 158681429 on Build GRCh38
UniGene ID:
Hs.407580
Species:
Human
siRNA ID
HSS112404
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PKP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005476.2
NP_001005476.1
14
2633
NM_001304969.1
NP_001291898.1
14
2630
NM_001304970.1
NP_001291899.1
14
2553
NM_001304971.1
NP_001291900.1
14
2633
NM_003628.4
NP_003619.2
14
2633
XM_011512016.1
XP_011510318.1
14
3070
XM_011512017.1
XP_011510319.1
14
2374
XM_011512018.1
XP_011510320.1
14
3064
XM_011512019.1
XP_011510321.1
14
2371
XM_011512020.2
XP_011510322.1
14
2417
XM_011512021.2
XP_011510323.1
14
2417
XM_011512022.1
XP_011510324.1
13
2585
XM_011512025.1
XP_011510327.1
9
1505
XM_017005122.1
XP_016860611.1
14
3067
XM_017005123.1
XP_016860612.1
14
3602
XM_017005124.1
XP_016860613.1
14
3061
XM_017005125.1
XP_016860614.1
14
2414
XM_017005126.1
XP_016860615.1
14
2374
XM_017005127.1
XP_016860616.1
14
3070
XM_017005128.1
XP_016860617.1
14
2414
XM_017005129.1
XP_016860618.1
13
2579
XM_017005130.1
XP_016860619.1
13
2582
XM_017005131.1
XP_016860620.1
9
1502
XM_017005132.1
XP_016860621.1
12
1910
XM_017005133.1
XP_016860622.1
13
2037
XM_017005134.1
XP_016860623.1
12
2404
XM_017005135.1
XP_016860624.1
11
1607
GenBank mRNA
AK054911.1
-
14
3070
AK124237.1
-
1
733
AK128720.1
-
12
2460
BC048013.1
-
14
2468
U60472.1
-
9
1847
X81889.1
-
14
2430
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell junction assembly
cell-cell signaling
single organismal cell-cell adhesion
regulation of cell adhesion
positive regulation of cytokinesis
positive regulation of GTPase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-tdxc8:80/100.66.76.99:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline