Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003630.2 | NP_003621.1 | 9 | 1034 | ||
GenBank mRNA | AB035307.1 | - | 9 | 918 | ||
AJ001625.1 | - | 9 | 835 | |||
AJ131389.1 | - | 9 | 831 | |||
AK025644.1 | - | 900 | ||||
AY277600.1 | - | 9 | 791 | |||
BC014551.1 | - | 9 | 972 | 4333011 | ||
BC015506.1 | - | 9 | 933 | 3914956 | ||
CR542062.1 | - | 9 | 772 |