Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003632.2 | NP_003623.1 | 21, 22 | 3784 | ||
XM_005257748.4 | XP_005257805.1 | 21, 22 | 4549 | |||
XM_017025238.1 | XP_016880727.1 | 23, 24 | 4919 | |||
GenBank mRNA | U87223.1 | - | 21, 22 | 3785 |