Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003648.2 | NP_003639.2 | 6, 7 | 770 | ||
NM_152879.2 | NP_690618.2 | 7, 8 | 835 | |||
XM_005246107.3 | XP_005246164.1 | 5, 6 | 502 | |||
XM_005246108.1 | XP_005246165.1 | 5, 6 | 656 | |||
XM_011512028.2 | XP_011510330.1 | 7, 8 | 2294 | |||
XM_011512029.2 | XP_011510331.1 | 7, 8 | 2295 | |||
XM_011512030.2 | XP_011510332.1 | 7, 8 | 891 | |||
XM_011512031.2 | XP_011510333.1 | 7, 8 | 765 | |||
XM_011512032.2 | XP_011510334.1 | 7, 8 | 749 | |||
XM_011512033.2 | XP_011510335.1 | 7, 8 | 2296 | |||
XM_011512035.1 | XP_011510337.1 | 5, 6 | 636 | |||
XM_011512037.2 | XP_011510339.1 | 5, 6 | 740 | |||
XM_011512039.2 | XP_011510341.1 | 7, 8 | 2296 | |||
XM_011512040.2 | XP_011510342.1 | 5, 6 | 672 | |||
XM_017005138.1 | XP_016860627.1 | 7, 8 | 1173 | |||
XM_017005139.1 | XP_016860628.1 | 5, 6 | 665 | |||
XM_017005140.1 | XP_016860629.1 | 7, 8 | 767 | |||
GenBank mRNA | AB078966.1 | - | 7, 8 | 835 | ||
AK308846.1 | - | 5, 6 | 656 | |||
AK311044.1 | - | 5, 6 | 589 | |||
D63479.3 | - | 7, 8 | 776 | |||
D73409.1 | - | 6, 7 | 771 |