Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003655.2 | NP_003646.2 | 2, 3 | 268 | ||
GenBank mRNA | AF013956.1 | - | 2, 3 | 271 | ||
AY390430.1 | - | 2, 3 | 105 | |||
BC014967.1 | - | 2, 3 | 124 | 4856728 | ||
BG029608.1 | - | 2, 3 | 101 | |||
BQ225831.1 | - | 268 | ||||
EU439707.1 | - | 2, 3 | 300 | |||
KU178422.1 | - | 2, 3 | 105 | |||
KU178423.1 | - | 2, 3 | 105 |