Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136540.1 | NP_001130012.1 | 6 | 568 | ||
NM_001136541.1 | NP_001130013.1 | 5 | 475 | |||
NM_003661.3 | NP_003652.2 | 6 | 529 | |||
NM_145343.2 | NP_663318.1 | 7 | 683 | |||
XM_005261796.3 | XP_005261853.1 | 5 | 454 | |||
XM_011530478.2 | XP_011528780.1 | 6 | 601 | |||
GenBank mRNA | AF019225.2 | - | 6 | 528 | ||
AF305224.1 | - | 6 | 468 | |||
AF305428.1 | - | 6 | 508 | |||
AF323540.1 | - | 7 | 682 | |||
AK296099.1 | - | 7 | 666 | |||
AK298053.1 | - | 3 | 259 | |||
AK300454.1 | - | 5 | 444 | |||
AK303919.1 | - | 6 | 502 | |||
AK309143.1 | - | 7 | 714 | |||
AK313752.1 | - | 7 | 622 | |||
BC017331.1 | - | 6 | 459 | |||
BC112943.1 | - | 6 | 484 | 4512933 | ||
BC113867.1 | - | 6 | 459 | |||
BC114475.1 | - | 6 | 459 | |||
BC127186.1 | - | 6 | 459 | 40034503 | ||
BC141823.1 | - | 6 | 484 | 40148156 | ||
BC142720.1 | - | 6 | 484 | |||
BC143038.1 | - | 6 | 459 | 40034505 | ||
BC143039.1 | - | 6 | 459 | 40034506 | ||
BG419709.1 | - | 5 | 398 | |||
DC340792.1 | - | 6 | 506 | |||
DC367266.1 | - | 5 | 475 | |||
KU178424.1 | - | 5 | 361 | |||
KU178425.1 | - | 4 | 307 |