Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271769.1 | NP_001258698.1 | 7 | 878 | ||
NM_003664.4 | NP_003655.3 | 7 | 866 | |||
XM_005248618.3 | XP_005248675.1 | 7 | 838 | |||
XM_005248619.4 | XP_005248676.1 | 7 | 838 | |||
XM_017010001.1 | XP_016865490.1 | 7 | 618 | |||
GenBank mRNA | AK225908.1 | - | 7 | 742 | ||
AK291201.1 | - | 7 | 807 | |||
AK303300.1 | - | 5 | 486 | |||
AY623424.1 | - | 7 | 768 | |||
BC038444.1 | - | 7 | 827 | 3914400 | ||
BX538041.1 | - | 7 | 816 | |||
KU178426.1 | - | 7 | 690 | |||
KU178427.1 | - | 7 | 690 | |||
U81504.1 | - | 7 | 782 | |||
U91931.1 | - | 7 | 743 |