Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003695.2 | NP_003686.1 | 2 | 175 | ||
GenBank mRNA | AK312164.1 | - | 2 | 122 | ||
BC022806.1 | - | 2 | 102 | |||
BC031330.1 | - | 2 | 175 | 5428818 | ||
BC034542.1 | - | 1 | 664 | |||
BM019592.1 | - | 2 | 186 | |||
X82693.1 | - | 2 | 110 |