Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173628.3 | NP_775899.3 | 69 | 11183 | ||
XM_011525416.2 | XP_011523718.1 | 69 | 11219 | |||
XM_017025261.1 | XP_016880750.1 | 27 | 4443 | |||
GenBank mRNA | AB290170.1 | - | 12 | 1884 | ||
AJ000522.1 | - | 8 | 1210 | |||
AK126059.1 | - | 1 | 720 | |||
AK128065.1 | - | 16 | 2720 | |||
AL122077.1 | - | 10 | 1506 | |||
AL832652.1 | - | 14 | 2254 |