Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASET2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003730.4 | NP_003721.2 | 4 | 653 | ||
XM_017011397.1 | XP_016866886.1 | 5 | 432 | |||
XM_017011398.1 | XP_016866887.1 | 3 | 554 | |||
XM_017011399.1 | XP_016866888.1 | 4 | 614 | |||
GenBank mRNA | AJ419865.1 | - | 4 | 607 | ||
AJ419866.1 | - | 4 | 607 | |||
AJ419867.1 | - | 4 | 607 | |||
AK315467.1 | - | 4 | 614 | |||
BC001660.2 | - | 4 | 579 | 2959583 | ||
BC001819.2 | - | 4 | 579 | 2959583 | ||
BC036282.1 | - | 4 | 640 | |||
BC039713.1 | - | 4 | 582 | 5225327 | ||
BC051912.1 | - | 4 | 592 | 5227463 | ||
U85625.2 | - | 4 | 610 |