Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277731.1 | NP_001264660.1 | 1 | 1539 | ||
NM_003734.3 | NP_003725.1 | 1 | 1539 | |||
NR_102422.1 | 1 | 1539 | ||||
XM_011525419.1 | XP_011523721.1 | 1 | 1600 | |||
XM_011525420.2 | XP_011523722.1 | 1 | 1600 | |||
GenBank mRNA | AF067406.1 | - | 1 | 1459 | ||
AK315129.1 | - | 1 | 1518 | |||
BC050549.1 | - | 1 | 1489 | 6193046 | ||
JX020506.1 | - | 1 | 1467 | |||
U39447.1 | - | 1 | 1539 |