Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243736.1 | NP_001230665.1 | 5 | 364 | ||
NM_021970.3 | NP_068805.1 | 5 | 385 | |||
NR_024170.1 | 4 | 339 | ||||
GenBank mRNA | AF201947.1 | - | 5 | 295 | ||
AK311755.1 | - | 5 | 246 | |||
AL544981.3 | - | 5 | 194 | |||
BC110402.1 | - | 4 | 205 | |||
BI562630.1 | - | 4 | 342 |