Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003747.2 | NP_003738.2 | 21 | 3195 | ||
XM_006716263.3 | XP_006716326.1 | 21 | 3901 | |||
XM_011543845.2 | XP_011542147.1 | 21 | 3901 | |||
XM_011543846.2 | XP_011542148.1 | 21 | 3901 | |||
XM_011543847.2 | XP_011542149.1 | 19 | 3588 | |||
GenBank mRNA | AB209339.1 | - | 20 | 2375 | ||
AF082556.1 | - | 21 | 3195 | |||
AF082557.1 | - | 20 | 2491 | |||
AK302534.1 | - | 21 | 2869 | |||
BC098394.1 | - | 21 | 3218 | 4374347 |