Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161504.1 | NP_001154976.1 | 4 | 408 | ||
NM_001319218.1 | NP_001306147.1 | 4 | 548 | |||
NM_003748.3 | NP_003739.2 | 4 | 548 | |||
NM_170726.2 | NP_733844.1 | 4 | 548 | |||
GenBank mRNA | AK222486.1 | - | 4 | 304 | ||
AK289972.1 | - | 4 | 474 | |||
AK294552.1 | - | 4 | 408 | |||
BC007581.1 | - | 4 | 278 | 3139944 | ||
BC023600.2 | - | 4 | 289 | 4548787 | ||
BM856369.1 | - | 4 | 327 | |||
DC318624.1 | - | 4 | 548 | |||
DC342814.1 | - | 4 | 548 | |||
FJ462711.1 | - | 4 | 286 | |||
U24266.1 | - | 4 | 302 | |||
U24267.1 | - | 4 | 302 |