Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098484.2 | NP_001091954.1 | 12 | 1532 | ||
NM_001134742.1 | NP_001128214.1 | 12 | 1532 | |||
NM_003759.3 | NP_003750.1 | 9 | 1479 | |||
XM_011532390.2 | XP_011530692.1 | 7 | 1018 | |||
XM_017008792.1 | XP_016864281.1 | 10 | 1205 | |||
XM_017008793.1 | XP_016864282.1 | 7 | 1504 | |||
GenBank mRNA | AB470072.1 | - | 11 | 1422 | ||
AF004813.1 | - | 10 | 1355 | |||
AF007216.1 | - | 9 | 1432 | |||
AF011390.1 | - | 12 | 1532 | |||
AF053753.1 | - | 11 | 1415 | |||
AF053754.1 | - | 11 | 1415 | |||
AF069510.1 | - | 11 | 1459 | |||
AF157492.1 | - | 12 | 1491 | |||
AF310248.1 | - | 11 | 1415 | |||
AK226164.1 | - | 10 | 1305 | |||
AK307594.1 | - | 12 | 1512 | |||
AK308038.1 | - | 10 | 1006 | |||
BC030977.1 | - | 9 | 1479 | 4610968 | ||
EF531618.1 | - | 11 | 1415 |