Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098484.2 | NP_001091954.1 | 14 | 1893 | ||
NM_001134742.1 | NP_001128214.1 | 14 | 1893 | |||
NM_003759.3 | NP_003750.1 | 11 | 1840 | |||
XM_011532390.2 | XP_011530692.1 | 9 | 1379 | |||
XM_017008792.1 | XP_016864281.1 | 12 | 1566 | |||
XM_017008793.1 | XP_016864282.1 | 9 | 1865 | |||
GenBank mRNA | AB470072.1 | - | 13 | 1783 | ||
AF004813.1 | - | 12 | 1716 | |||
AF007216.1 | - | 11 | 1793 | |||
AF011390.1 | - | 14 | 1893 | |||
AF053753.1 | - | 13 | 1776 | |||
AF053754.1 | - | 13 | 1776 | |||
AF069510.1 | - | 13 | 1820 | |||
AF157492.1 | - | 14 | 1852 | |||
AF310248.1 | - | 13 | 1776 | |||
AK226164.1 | - | 12 | 1666 | |||
AK307594.1 | - | 14 | 1873 | |||
AK308038.1 | - | 12 | 1368 | |||
BC030977.1 | - | 11 | 1840 | 4610968 | ||
EF531618.1 | - | 13 | 1776 |