Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001433.2 | NP_001001433.1 | 6 | 1328 | ||
NM_001134772.2 | NP_001128244.1 | 5 | 1316 | |||
NM_001134773.2 | NP_001128245.1 | 6 | 1277 | |||
NM_001204868.1 | NP_001191797.1 | 7 | 690 | |||
NM_003763.5 | NP_003754.2 | 5 | 1265 | |||
NR_037941.1 | 5 | 1114 | ||||
NR_037942.1 | 6 | 507 | ||||
NR_037943.1 | 5 | 464 | ||||
GenBank mRNA | AF008935.1 | - | 5 | 648 | ||
AF008936.1 | - | 6 | 711 | |||
AF008937.1 | - | 5 | 500 | |||
AF038897.1 | - | 6 | 520 | |||
AF428146.1 | - | 6 | 523 | |||
AK026970.1 | - | 6 | 459 | |||
AK296282.1 | - | 3 | 227 | |||
AK304867.1 | - | 6 | 937 | |||
AK316547.1 | - | 6 | 507 | |||
BC019042.2 | - | 5 | 720 | 4585425 | ||
BC073876.1 | - | 5 | 627 | 4371823 | ||
BQ229292.1 | - | 5 | 413 | |||
BU844520.1 | - | 6 | 475 | |||
BX396221.2 | - | 6 | 645 | |||
DC424825.1 | - | 6 | 507 |
Gene Symbol: STX16-NPEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037945.1 | 6 | 1328 | |||
GenBank mRNA | AB209296.1 | - | 6 | 1147 |