Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297576.1 | NP_001284505.1 | 5 | 700 | ||
NM_001297577.1 | NP_001284506.1 | 4 | 498 | |||
NM_001297578.1 | NP_001284507.1 | 3 | 327 | |||
NM_003768.4 | NP_003759.1 | 3 | 393 | |||
NR_123724.1 | 2 | 219 | ||||
XM_006711599.2 | XP_006711662.1 | 3 | 297 | |||
GenBank mRNA | AK095879.1 | - | 5 | 605 | ||
AK311762.1 | - | 3 | 322 | |||
BC002426.2 | - | 3 | 269 | 3346270 | ||
BC022554.1 | - | 3 | 380 | 4820225 | ||
BT007252.1 | - | 2 | 187 | |||
CR541733.1 | - | 2 | 187 | |||
DA131259.1 | - | 3 | 377 | |||
DA157938.1 | - | 3 | 263 | |||
DA532423.1 | - | 3 | 377 | |||
DA538845.1 | - | 2 | 157 | |||
DA707281.1 | - | 4 | 425 | |||
HY182028.1 | - | 3 | 396 | |||
Y13736.1 | - | 3 | 256 |