Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JRKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261833.1 | NP_001248762.1 | 2 | 1405 | ||
NM_003772.3 | NP_003763.2 | 1 | 1814 | |||
GenBank mRNA | AF004715.1 | - | 1 | 1118 | ||
AK290579.1 | - | 2 | 1405 | |||
AK314216.1 | - | 1 | 1503 | |||
BC109202.2 | - | 1 | 1158 | 40005971 |