Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004661.3 | NP_004652.2 | 14 | 1497 | ||
GenBank mRNA | AB011472.1 | - | 14 | 1484 | ||
AF053977.1 | - | 14 | 1464 | |||
AF191341.1 | - | 14 | 1448 | |||
AK223250.1 | - | 14 | 1485 | |||
AK291610.1 | - | 14 | 1479 | |||
AK304635.1 | - | 13 | 1361 | |||
BC017713.1 | - | 14 | 1456 | 4399056 | ||
BT009810.1 | - | 14 | 1448 |