Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003778.3 | NP_003769.1 | 6, 7 | 1220 | ||
NM_212543.1 | NP_997708.1 | 7, 8 | 1303 | |||
XM_005247855.1 | XP_005247912.1 | 6, 7 | 1278 | |||
XM_006713798.2 | XP_006713861.1 | 7, 8 | 1363 | |||
XM_006713799.2 | XP_006713862.1 | 7, 8 | 1408 | |||
XM_006713800.1 | XP_006713863.1 | 6, 7 | 1255 | |||
XM_006713801.2 | XP_006713864.1 | 8, 9 | 1446 | |||
XM_011513260.1 | XP_011511562.1 | 8, 9 | 1375 | |||
GenBank mRNA | AB024436.1 | - | 4, 5 | 804 | ||
AF020920.1 | - | 5, 6 | 1111 | |||
AF022367.1 | - | 4, 5 | 801 | |||
AF038662.1 | - | 5, 6 | 1111 | |||
AK001006.1 | - | 7, 8 | 1276 | |||
AK314425.1 | - | 6, 7 | 1193 | |||
AY359008.1 | - | 4, 5 | 919 | |||
BC004523.2 | - | 5, 6 | 969 | 3531796 | ||
BC062618.1 | - | 6, 7 | 1442 | 5189057 | ||
BX647468.1 | - | 6, 7 | 1193 | |||
CR749555.1 | - | 6, 7 | 1203 |