Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003778.3 | NP_003769.1 | 7 | 1260 | ||
NM_212543.1 | NP_997708.1 | 8 | 1343 | |||
XM_005247855.1 | XP_005247912.1 | 7 | 1318 | |||
XM_006713798.2 | XP_006713861.1 | 8 | 1403 | |||
XM_006713799.2 | XP_006713862.1 | 8 | 1448 | |||
XM_006713800.1 | XP_006713863.1 | 7 | 1295 | |||
XM_006713801.2 | XP_006713864.1 | 9 | 1486 | |||
XM_011513260.1 | XP_011511562.1 | 9 | 1415 | |||
XM_017007401.1 | XP_016862890.1 | 6 | 1110 | |||
GenBank mRNA | AB024436.1 | - | 5 | 844 | ||
AF020920.1 | - | 6 | 1151 | |||
AF022367.1 | - | 5 | 841 | |||
AF038662.1 | - | 6 | 1151 | |||
AK001006.1 | - | 8 | 1316 | |||
AK314425.1 | - | 7 | 1233 | |||
AY359008.1 | - | 5 | 959 | |||
BC004523.2 | - | 6 | 1009 | 3531796 | ||
BC062618.1 | - | 7 | 1482 | 5189057 | ||
BX647468.1 | - | 7 | 1233 | |||
CR749555.1 | - | 7 | 1243 |