Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003778.3 | NP_003769.1 | 8 | 1372 | ||
NM_212543.1 | NP_997708.1 | 9 | 1455 | |||
XM_005247855.1 | XP_005247912.1 | 8 | 1430 | |||
XM_006713798.2 | XP_006713861.1 | 9 | 1515 | |||
XM_006713799.2 | XP_006713862.1 | 9 | 1560 | |||
XM_006713800.1 | XP_006713863.1 | 8 | 1407 | |||
XM_006713801.2 | XP_006713864.1 | 10 | 1598 | |||
XM_011513260.1 | XP_011511562.1 | 10 | 1527 | |||
XM_017007401.1 | XP_016862890.1 | 7 | 1222 | |||
GenBank mRNA | AB024436.1 | - | 6 | 956 | ||
AF020920.1 | - | 7 | 1263 | |||
AF022367.1 | - | 6 | 953 | |||
AF038662.1 | - | 7 | 1263 | |||
AK001006.1 | - | 9 | 1428 | |||
AK123182.1 | - | 1 | 935 | |||
AK314425.1 | - | 8 | 1345 | |||
AL833580.1 | - | 1 | 3453 | |||
AY359008.1 | - | 6 | 1071 | |||
BC004523.2 | - | 7 | 1121 | 3531796 | ||
BC062618.1 | - | 8 | 1594 | 5189057 | ||
BX647468.1 | - | 7 | 4903 | |||
CR749555.1 | - | 8 | 1355 |