Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003783.3 | NP_003774.1 | 2 | 739 | ||
GenBank mRNA | AF288390.1 | - | 2 | 719 | ||
AK314116.1 | - | 2 | 457 | |||
BC022507.1 | - | 2 | 739 | 4792498 | ||
Y15060.1 | - | 1 | 24 |