Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOL4 SIRNA Assays ›
Gene Symbol:
NOL4
Gene Name:
nucleolar protein 4
Gene Aliases:
CT125, HRIHFB2255, NOLP
Chromosome Location:
Chr.18: 33851100 - 34224834 on Build GRCh38
UniGene ID:
Hs.514795
Species:
Human
siRNA ID
HSS112804
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NOL4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198546.1
NP_001185475.1
6
1092
NM_001198547.1
NP_001185476.1
6
981
NM_001198548.1
NP_001185477.1
6
1092
NM_001198549.1
NP_001185478.1
2
196
NM_001282527.1
NP_001269456.1
7
1022
NM_003787.4
NP_003778.2
6
1092
XM_006722563.2
XP_006722626.1
7
1849
XM_006722564.2
XP_006722627.1
6
834
XM_006722565.1
XP_006722628.1
7
868
XM_011526237.1
XP_011524539.1
7
1846
XM_011526238.1
XP_011524540.1
6
1458
XM_011526239.2
XP_011524541.1
5
850
XM_017026052.1
XP_016881541.1
5
680
XM_017026053.1
XP_016881542.1
6
832
XM_017026054.1
XP_016881543.1
7
960
XM_017026055.1
XP_016881544.1
7
1499
XM_017026056.1
XP_016881545.1
7
753
XM_017026057.1
XP_016881546.1
7
1424
XM_017026058.1
XP_016881547.1
6
571
XM_017026059.1
XP_016881548.1
7
2439
XM_017026060.1
XP_016881549.1
2
210
GenBank mRNA
AB017800.1
-
6
1102
AK091463.1
-
7
871
AK296539.1
-
2
179
AK297185.1
-
6
746
AK299850.1
-
6
603
AK312562.1
-
7
619
BC000313.2
-
6
778
2821116
BT006763.1
-
5
452
CR456730.1
-
5
452
DC350556.1
-
2
196
Back To Top
More Information
Gene Ontology Categories:
Process(es)
RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-nbn7j:80/100.66.77.222:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline