Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252641.1 | NP_001239570.1 | 10 | 1231 | ||
NM_003796.3 | NP_003787.2 | 10 | 1546 | |||
NR_045557.1 | 9 | 1511 | ||||
XM_005259362.2 | XP_005259419.1 | 10 | 1225 | |||
XM_005259363.4 | XP_005259420.1 | 10 | 1282 | |||
XM_011527435.1 | XP_011525737.1 | 9 | 1108 | |||
GenBank mRNA | AF091095.1 | - | 11 | 1179 | ||
AK001437.1 | - | 9 | 1229 | |||
AK226173.1 | - | 10 | 1531 | |||
AK292170.1 | - | 10 | 1279 | |||
AK314313.1 | - | 10 | 1268 | |||
AW954529.1 | - | 4 | 668 | |||
BC014933.2 | - | 5 | 760 | |||
BC026184.2 | - | 10 | 1228 | |||
BC067259.1 | - | 9 | 1235 | 6500727 |