Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNGTT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286426.1 | NP_001273355.1 | 2 | 361 | ||
NM_003800.4 | NP_003791.3 | 2 | 361 | |||
XM_017011401.1 | XP_016866890.1 | 2 | 448 | |||
XM_017011402.1 | XP_016866891.1 | 2 | 448 | |||
GenBank mRNA | AB009022.1 | - | 2 | 141 | ||
AB009023.1 | - | 2 | 141 | |||
AB009024.1 | - | 2 | 141 | |||
AB012142.1 | - | 2 | 296 | |||
AB012143.1 | - | 2 | 296 | |||
AF025654.1 | - | 2 | 429 | |||
AK295719.1 | - | 3 | 239 | |||
AK312407.1 | - | 2 | 332 | |||
BC019954.1 | - | 2 | 328 | 3923156 | ||
BP256840.1 | - | 2 | 361 |