Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135653.1 | NP_001129125.1 | 3 | 1323 | ||
NM_001135654.1 | NP_001129126.1 | 3 | 1323 | |||
NM_003819.3 | NP_003810.1 | 3 | 1323 | |||
GenBank mRNA | BC029969.1 | - | 3 | 875 | ||
BC065540.1 | - | 3 | 1226 | 6061561 | ||
BC071591.1 | - | 3 | 1261 | 5265829 | ||
BC094755.1 | - | 3 | 1264 | 30331896 | ||
U33818.1 | - | 3 | 578 | |||
U75686.1 | - | 3 | 1219 |