Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003820.3 | NP_003811.2 | 6 | 907 | ||
XM_011542383.2 | XP_011540685.1 | 3 | 950 | |||
XM_017002719.1 | XP_016858208.1 | 3 | 1171 | |||
XM_017002720.1 | XP_016858209.1 | 6 | 1625 | |||
XM_017002721.1 | XP_016858210.1 | 7 | 1035 | |||
GenBank mRNA | AB208808.1 | - | 5 | 2234 | ||
AF153978.1 | - | 6 | 852 | |||
AF373877.1 | - | 6 | 607 | |||
AF373878.1 | - | 6 | 607 | |||
AK124010.1 | - | 4 | 1806 | |||
AY358879.1 | - | 6 | 688 | |||
BC002794.2 | - | 6 | 873 | 3614650 | ||
BC018882.2 | - | 6 | 873 | |||
BC029848.1 | - | 4 | 844 | 5178561 | ||
CR456909.1 | - | 6 | 607 | |||
U70321.1 | - | 6 | 900 | |||
U81232.1 | - | 6 | 871 |