Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003827.3 | NP_003818.2 | 10, 11 | 1101 | ||
NR_038456.1 | 10, 11 | 1101 | ||||
NR_038457.1 | 8, 9 | 904 | ||||
XM_011527436.1 | XP_011525738.1 | 10, 11 | 775 | |||
XM_011527437.1 | XP_011525739.1 | 9, 10 | 854 | |||
GenBank mRNA | AK098401.1 | - | 10, 11 | 934 | ||
AK126519.1 | - | 4, 5 | 3143 | |||
AK292244.1 | - | 10, 11 | 865 | |||
AW206113.1 | - | 2, 3 | 425 | |||
BC001165.1 | - | 10, 11 | 873 | 3533668 | ||
BC007432.2 | - | 10, 11 | 864 | 2962847 | ||
BC028234.1 | - | 10, 11 | 919 | 5245770 | ||
BC091511.1 | - | 10, 11 | 880 | 30512911 | ||
BT019568.1 | - | 10, 11 | 783 | |||
BU168063.1 | - | 5, 6 | 339 | |||
CX873074.1 | - | 8, 9 | 646 | |||
U39412.1 | - | 10, 11 | 850 |