Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003849.3 | NP_003840.2 | 3 | 485 | ||
GenBank mRNA | AF104921.2 | - | 3 | 284 | ||
AK225737.1 | - | 3 | 262 | |||
AK291082.1 | - | 3 | 284 | |||
AK296741.1 | - | 3 | 292 | |||
AK313290.1 | - | 3 | 264 | |||
BC000504.2 | - | 3 | 288 | 2821634 | ||
BM804835.1 | - | 3 | 471 | |||
CR457139.1 | - | 3 | 253 | |||
Z68204.1 | - | 3 | 259 |