Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317034.1 | NP_001303963.1 | 8 | 681 | ||
NM_001317035.1 | NP_001303964.1 | 8 | 657 | |||
NM_003859.2 | NP_003850.1 | 7 | 576 | |||
NR_133648.1 | 7 | 598 | ||||
XM_011529093.2 | XP_011527395.1 | 7 | 599 | |||
GenBank mRNA | AK289569.1 | - | 7 | 544 | ||
BC007073.1 | - | 7 | 562 | 3997453 | ||
BC008466.1 | - | 7 | 562 | 4276338 | ||
BC016322.1 | - | 7 | 547 | 4083041 | ||
BM922866.1 | - | 8 | 673 | |||
CA865473.1 | - | 3 | 533 | |||
CR456926.1 | - | 7 | 535 | |||
D86198.1 | - | 7 | 535 | |||
DR156925.1 | - | 8 | 636 | |||
KU178444.1 | - | 7 | 535 | |||
KU178445.1 | - | 8 | 616 | |||
KU178446.1 | - | 7 | 573 |