Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282236.1 | NP_001269165.1 | 9 | 1742 | ||
NM_004196.4 | NP_004187.2 | 8 | 890 | |||
XM_005268157.2 | XP_005268214.1 | 9 | 1482 | |||
XM_005268160.4 | XP_005268217.1 | 6 | 2578 | |||
XM_011537276.1 | XP_011535578.1 | 9 | 892 | |||
XM_017021729.1 | XP_016877218.1 | 9 | 1374 | |||
XM_017021732.1 | XP_016877221.1 | 9 | 1461 | |||
GenBank mRNA | AA780354.1 | - | 1 | 128 | ||
AK308732.1 | - | 9 | 1737 | |||
BC104977.1 | - | 8 | 865 | 8143980 | ||
X66358.1 | - | 8 | 1050 |