Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003864.3 | NP_003855.1 | 1, 2 | 567 | ||
GenBank mRNA | AF055993.1 | - | 1, 2 | 566 | ||
BC016757.1 | - | 1, 2 | 562 | 4074154 |