Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004664.3 | NP_004655.1 | 2, 3 | 393 | ||
NR_136888.1 | 2, 3 | 393 | ||||
XM_011538928.2 | XP_011537230.1 | 2, 3 | 387 | |||
GenBank mRNA | AF028826.1 | - | 1, 2 | 109 | ||
AF087693.1 | - | 2, 3 | 362 | |||
AF173081.1 | - | 2, 3 | 393 | |||
AK309256.1 | - | 2, 3 | 182 | |||
AK315321.1 | - | 2, 3 | 237 | |||
BC062332.1 | - | 2, 3 | 258 | |||
BC099921.1 | - | 2, 3 | 203 | 6729473 | ||
BC118609.1 | - | 2, 3 | 337 | 40109202 | ||
BC122561.1 | - | 2, 3 | 337 | 40109204 | ||
CR407680.1 | - | 2, 3 | 200 |