Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184879.1 | NP_001171808.1 | 7 | 1013 | ||
NM_001184881.1 | NP_001171810.1 | 5 | 864 | |||
NM_001184882.1 | NP_001171811.1 | 5 | 620 | |||
NM_003874.3 | NP_003865.1 | 6 | 962 | |||
XM_011510094.2 | XP_011508396.1 | 7 | 976 | |||
XM_011510095.2 | XP_011508397.1 | 6 | 652 | |||
GenBank mRNA | AF054815.1 | - | 6 | 952 | ||
AF054816.1 | - | 7 | 1003 | |||
AF054817.1 | - | 5 | 854 | |||
AJ223324.1 | - | 6 | 927 | |||
AK296290.1 | - | 5 | 581 | |||
AK313496.1 | - | 6 | 928 | |||
BC020063.1 | - | 6 | 907 | 4385965 | ||
CR933666.1 | - | 5 | 581 | |||
U82988.1 | - | 6 | 924 | |||
Y12632.1 | - | 7 | 985 |