Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003883.3 | NP_003874.2 | 5, 6 | 478 | ||
XM_011537697.2 | XP_011535999.1 | 4, 5 | 330 | |||
GenBank mRNA | AF005482.1 | - | 3, 4 | 494 | ||
AF039703.1 | - | 5, 6 | 434 | |||
AY429538.1 | - | 3, 4 | 161 | |||
BC000614.1 | - | 5, 6 | 467 | 3345755 | ||
BM461585.1 | - | 5, 6 | 468 | |||
U66914.1 | - | 5, 6 | 423 | |||
U75696.1 | - | 4, 5 | 461 | |||
U75697.1 | - | 5, 6 | 467 |