Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003887.2 | NP_003878.1 | 23, 24 | 2846 | ||
XM_006711898.1 | XP_006711961.1 | 24, 25 | 2608 | |||
XM_006711901.3 | XP_006711964.1 | 24, 25 | 2458 | |||
XM_011510403.2 | XP_011508705.1 | 24, 25 | 2629 | |||
XM_011510404.2 | XP_011508706.1 | 23, 24 | 2619 | |||
XM_011510405.2 | XP_011508707.1 | 23, 24 | 2555 | |||
XM_011510406.2 | XP_011508708.1 | 25, 26 | 2562 | |||
XM_011510409.1 | XP_011508711.1 | 24, 25 | 2500 | |||
GenBank mRNA | AB007860.1 | - | 23, 24 | 2846 |