Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003889.3 | NP_003880.3 | 2 | 1997 | ||
NM_022002.2 | NP_071285.1 | 2 | 323 | |||
NM_033013.2 | NP_148934.1 | 2 | 1997 | |||
GenBank mRNA | AB307701.1 | - | 1 | 158 | ||
AF061056.1 | - | 2 | 461 | |||
AF084644.1 | - | 2 | 334 | |||
AF084645.1 | - | 2 | 437 | |||
AJ009936.1 | - | 3 | 1997 | |||
AJ009937.1 | - | 3 | 1997 | |||
AK223456.1 | - | 2 | 442 | |||
AK310789.1 | - | 2 | 446 | |||
AY091855.1 | - | 2 | 740 | |||
BC017304.2 | - | 2 | 397 | 5087401 | ||
HQ692837.1 | - | 2 | 275 | |||
HQ709177.1 | - | 2 | 275 |