Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003889.3 | NP_003880.3 | 4 | 2331 | ||
NM_022002.2 | NP_071285.1 | 4 | 657 | |||
NM_033013.2 | NP_148934.1 | 4 | 2331 | |||
GenBank mRNA | AB307701.1 | - | 3 | 492 | ||
AF061056.1 | - | 4 | 795 | |||
AF084644.1 | - | 4 | 668 | |||
AF084645.1 | - | 4 | 771 | |||
AJ009936.1 | - | 5 | 2331 | |||
AJ009937.1 | - | 5 | 2331 | |||
AK122990.1 | - | 1 | 624 | |||
AK223456.1 | - | 4 | 776 | |||
AY091855.1 | - | 4 | 1074 | |||
BC017304.2 | - | 4 | 731 | 5087401 | ||
HQ692837.1 | - | 4 | 609 | |||
HQ709177.1 | - | 4 | 600 |