Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178088.1 | NP_001171559.1 | 2, 3 | 515 | ||
NM_003898.3 | NP_003889.1 | 3, 4 | 592 | |||
XM_005267198.4 | XP_005267255.1 | 2, 3 | 339 | |||
XM_005267199.2 | XP_005267256.1 | 2, 3 | 339 | |||
XM_006715592.3 | XP_006715655.1 | 3, 4 | 592 | |||
XM_011536224.1 | XP_011534526.1 | 3, 4 | 592 | |||
XM_011536225.1 | XP_011534527.1 | 3, 4 | 632 | |||
XM_011536227.2 | XP_011534529.1 | 3, 4 | 592 | |||
XM_011536230.2 | XP_011534532.1 | 3, 4 | 592 | |||
XM_017011406.1 | XP_016866895.1 | 2, 3 | 339 | |||
GenBank mRNA | AB002346.2 | - | 2, 3 | 328 | ||
AF039945.1 | - | 2, 3 | 328 | |||
AF318616.1 | - | 3, 4 | 569 | |||
AK294480.1 | - | 3, 4 | 592 | |||
AK296242.1 | - | 2, 3 | 603 | |||
AK309018.1 | - | 2, 3 | 603 | |||
AY152396.1 | - | 3, 4 | 494 | |||
BX648749.1 | - | 2, 3 | 422 | |||
DA327619.1 | - | 2, 3 | 515 |