Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242350.2 | NP_001229279.1 | 2 | 444 | ||
NM_004665.4 | NP_004656.2 | 2 | 444 | |||
NM_078488.2 | NP_511043.1 | 3 | 272 | |||
NR_034173.2 | 2 | 444 | ||||
NR_034174.2 | 2 | 444 | ||||
NR_110143.1 | 2 | 444 | ||||
NR_110144.1 | 2 | 444 | ||||
NR_110145.1 | 2 | 444 | ||||
NR_110146.1 | 2 | 444 | ||||
XM_006715593.3 | XP_006715656.1 | 4 | 419 | |||
XM_011536231.1 | XP_011534533.1 | 2 | 340 | |||
XM_017011407.1 | XP_016866896.1 | 3 | 1055 | |||
XM_017011408.1 | XP_016866897.1 | 3 | 302 | |||
XM_017011409.1 | XP_016866898.1 | 3 | 243 | |||
GenBank mRNA | AB026705.1 | - | 2 | 326 | ||
AB435062.1 | - | 2 | 319 | |||
AB435063.1 | - | 2 | 319 | |||
AB435064.1 | - | 2 | 319 | |||
AB435065.1 | - | 2 | 319 | |||
AJ132100.1 | - | 2 | 330 | |||
AK290908.1 | - | 2 | 340 | |||
AK292125.1 | - | 3 | 260 | |||
AK298719.1 | - | 2 | 340 | |||
BC126145.1 | - | 2 | 406 | 8991861 | ||
BC126147.1 | - | 2 | 406 | 8991863 | ||
D89974.1 | - | 2 | 330 | |||
DC412933.1 | - | 3 | 415 | |||
DC428640.1 | - | 2 | 444 | |||
DC428943.1 | - | 2 | 338 |